ADVH49B82568 AChemBlock

trans-octahydro-1H-pyrrolo[3,4-b]pyridine dihydrochloride

  • Molecular Weight : 199.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/trans-octahydro-1h-pyrrolo-3-4-b-pyridine-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84348
Availability details for
Quantity
  • Availability
  • Add to Cart