ADVH49B82569 AChemBlock

3-[(2R)-morpholin-2-yl]propanoic acid hydrochloride

  • Cas Number : 1793064-24-0
  • Molecular Weight : 195.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-2r-morpholin-2-yl-propanoic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78293
Availability details for
Quantity
  • Availability
  • Add to Cart