ADVH49B8256A AChemBlock

trans-6-methyl-octahydropyrrolo[3,4-b][1,4]oxazine

  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/trans-6-methyl-octahydropyrrolo-3-4-b-1-4-oxazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84350
Availability details for
Quantity
  • Availability
  • Add to Cart