ADVH49B8256B AChemBlock

NCB-0846

  • Cas Number : 1792999-26-8
  • Molecular Weight : 375.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/ncb-0846.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72371
Availability details for
Quantity
  • Availability
  • Add to Cart