ADVH49B8256F AChemBlock

5-methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridin-2-amine

  • Cas Number : 17899-48-8
  • Molecular Weight : 169.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-methyl-4-5-6-7-tetrahydrothiazolo-5-4-c-pyridin-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71610
Availability details for
Quantity
  • Availability
  • Add to Cart