ADVH49B82572 AChemBlock

trans-3,4-difluorocyclopentane-1-carboxylic acid

  • Molecular Weight : 150.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/trans-3-4-difluorocyclopentane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84359
Availability details for
Quantity
  • Availability
  • Add to Cart