ADVH49B82573 AChemBlock

trans-2-methyloxetan-3-amine

  • Molecular Weight : 87.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/trans-2-methyloxetan-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84360
Availability details for
Quantity
  • Availability
  • Add to Cart