ADVH49B8257B AChemBlock

trans-1-[(tert-butoxy)carbonyl]-4-(2,2,2-trifluoroethyl)pyrrolidine-3-carboxylic acid

  • Molecular Weight : 297.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/trans-1-tert-butoxy-carbonyl-4-2-2-2-trifluoroethyl-pyrrolidine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84368
Availability details for
Quantity
  • Availability
  • Add to Cart