ADVH49B8257D AChemBlock

tert-butyl N-[cis-3-hydroxypiperidin-4-yl]carbamate

  • Cas Number : 1787258-87-0
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-cis-3-hydroxypiperidin-4-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78312
Availability details for
Quantity
  • Availability
  • Add to Cart