ADVH49B8258A AChemBlock

tert-butyl octahydropyrrolo[3,4-b][1,4]oxazine-6-carboxylate hydrochloride

  • Molecular Weight : 264.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-octahydropyrrolo-3-4-b-1-4-oxazine-6-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84384
Availability details for
Quantity
  • Availability
  • Add to Cart