ADVH49B8258E AChemBlock

tert-butyl N-methyl-N-[(3R,5S)-5-methylpyrrolidin-3-yl]carbamate

  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-methyl-n-3r-5s-5-methylpyrrolidin-3-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84388
Availability details for
Quantity
  • Availability
  • Add to Cart