ADVH49B8258F AChemBlock

tert-butyl N-methyl-N-[(3R,5S)-5-(trifluoromethyl)piperidin-3-yl]carbamate

  • Molecular Weight : 282.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-methyl-n-3r-5s-5-trifluoromethyl-piperidin-3-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84389
Availability details for
Quantity
  • Availability
  • Add to Cart