ADVH49B82590 AChemBlock

tert-butyl n-{9-aminospiro[5.5]undecan-3-yl}carbamate

  • Molecular Weight : 282.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-9-aminospiro-5-5-undecan-3-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84390
Availability details for
Quantity
  • Availability
  • Add to Cart