ADVH49B82593 AChemBlock

tert-butyl N-{6,8-dioxo-5,7-diazaspiro[3.4]octan-2-yl}carbamate

  • Molecular Weight : 255.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-6-8-dioxo-5-7-diazaspiro-3-4-octan-2-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84393
Availability details for
Quantity
  • Availability
  • Add to Cart