ADVH49B8259F AChemBlock

tert-butyl N-{[5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-4-yl]methyl}carbamate

  • Molecular Weight : 335.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-5-tetramethyl-1-3-2-dioxaborolan-2-yl-pyrimidin-4-yl-methyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84405
Availability details for
Quantity
  • Availability
  • Add to Cart