ADVH49B825A3 AChemBlock

tert-butyl N-{[(3R)-1-(4-bromobenzoyl)piperidin-3-yl]methyl}carbamate

  • Molecular Weight : 397.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-3r-1-4-bromobenzoyl-piperidin-3-yl-methyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84409
Availability details for
Quantity
  • Availability
  • Add to Cart