ADVH49B825AB AChemBlock

tert-butyl N-{[(2R,4S)-4-(trifluoromethyl)pyrrolidin-2-yl]methyl}carbamate

  • Molecular Weight : 268.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-2r-4s-4-trifluoromethyl-pyrrolidin-2-yl-methyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84417
Availability details for
Quantity
  • Availability
  • Add to Cart