ADVH49B825B2 AChemBlock

tert-butyl N-[octahydro-1H-isoindol-4-yl]carbamate

  • Molecular Weight : 240.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-octahydro-1h-isoindol-4-yl-carbamate-307735.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84425
Availability details for
Quantity
  • Availability
  • Add to Cart