ADVH49B825B4 AChemBlock

tert-butyl N-[7-(hydroxymethyl)spiro[3.5]nonan-2-yl]carbamate

  • Molecular Weight : 269.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-7-hydroxymethyl-spiro-3-5-nonan-2-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84429
Availability details for
Quantity
  • Availability
  • Add to Cart