ADVH49B825B5 AChemBlock

tert-butyl N-[6-hydroxy-6-(trifluoromethyl)spiro[3.3]heptan-2-yl]carbamate

  • Molecular Weight : 295.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-6-hydroxy-6-trifluoromethyl-spiro-3-3-heptan-2-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84430
Availability details for
Quantity
  • Availability
  • Add to Cart