ADVH49B825B7 AChemBlock

tert-butyl N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridin-3-yl]carbamate

  • Molecular Weight : 359.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-5-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-pyrrolo-2-3-b-pyridin-3-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84432
Availability details for
Quantity
  • Availability
  • Add to Cart