ADVH49B825B9 AChemBlock

5,5-difluoro-2,3,3a,4,6,6a-hexahydro-1H-cyclopenta[c]pyrrole hydrochloride

  • Cas Number : 1782456-99-8
  • Molecular Weight : 183.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-5-difluoro-2-3-3a-4-6-6a-hexahydro-1h-cyclopenta-c-pyrrole-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78391
Availability details for
Quantity
  • Availability
  • Add to Cart