ADVH49B825BC AChemBlock

tert-butyl N-[3-(difluoromethyl)-1-bicyclo[1.1.1]pentanyl]carbamate

  • Molecular Weight : 233.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-3-difluoromethyl-1-bicyclo-1-1-1-pentanyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84437
Availability details for
Quantity
  • Availability
  • Add to Cart