ADVH49B825BF AChemBlock

2,3,4,5-tetrahydro-1H-1-benzazepin-9-amine

  • Cas Number : 1780911-57-0
  • Molecular Weight : 162.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-3-4-5-tetrahydro-1h-1-benzazepin-9-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78440
Availability details for
Quantity
  • Availability
  • Add to Cart