ADVH49B825C1 AChemBlock

tert-butyl N-[2-hydroxy-1-(oxetan-3-yl)ethyl]carbamate

  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-2-hydroxy-1-oxetan-3-yl-ethyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84442
Availability details for
Quantity
  • Availability
  • Add to Cart