ADVH49B825C3 AChemBlock

tert-butyl N-[2-(4-bromo-1-methyl-pyrazol-3-yl)ethyl]-N-methyl-carbamate

  • Molecular Weight : 318.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-2-4-bromo-1-methyl-pyrazol-3-yl-ethyl-n-methyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84444
Availability details for
Quantity
  • Availability
  • Add to Cart