ADVH49B825C5 AChemBlock

tert-butyl N-[2-(2-hydroxyethyl)pyrimidin-4-yl]carbamate

  • Molecular Weight : 239.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-2-2-hydroxyethyl-pyrimidin-4-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84446
Availability details for
Quantity
  • Availability
  • Add to Cart