ADVH49B825C7 AChemBlock

9-amino-1,3,4,5-tetrahydro-1-benzazepin-2-one

  • Cas Number : 1780415-06-6
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/9-amino-1-3-4-5-tetrahydro-1-benzazepin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78460
Availability details for
Quantity
  • Availability
  • Add to Cart