ADVH49B825CA AChemBlock

trans-4-amino-3-(methoxy)tetrahydropyran hydrochloride

  • Cas Number : 1778734-55-6
  • Molecular Weight : 167.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/trans-4-amino-3-methoxy-tetrahydropyran-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78497
Availability details for
Quantity
  • Availability
  • Add to Cart