ADVH49B825CC AChemBlock

tert-butyl N-[(7S)-5-azaspiro[2.4]heptan-7-yl]-N-methylcarbamate

  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-7s-5-azaspiro-2-4-heptan-7-yl-n-methylcarbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84453
Availability details for
Quantity
  • Availability
  • Add to Cart