ADVH49B825CE AChemBlock

tert-butyl N-[(6R)-1,4-oxazepan-6-yl]carbamate

  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-6r-1-4-oxazepan-6-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84455
Availability details for
Quantity
  • Availability
  • Add to Cart