ADVH49B825CF AChemBlock

tert-butyl N-[(5-bromo-2-methyl-2H-indazol-3-yl)methyl]carbamate

  • Molecular Weight : 340.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-5-bromo-2-methyl-2h-indazol-3-yl-methyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84456
Availability details for
Quantity
  • Availability
  • Add to Cart