ADVH49B825D2 AChemBlock

tert-butyl (1S,4S)-3-oxo-2-thia-5-azabicyclo[2.2.1]heptane-5-carboxylate

  • Cas Number : 177615-42-8
  • Molecular Weight : 229.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1s-4s-3-oxo-2-thia-5-azabicyclo-2-2-1-heptane-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78504
Availability details for
Quantity
  • Availability
  • Add to Cart