ADVH49B825D4 AChemBlock

tert-butyl N-[(3S,4S)-4-methoxypiperidin-3-yl]carbamate

  • Molecular Weight : 230.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-3s-4s-4-methoxypiperidin-3-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84461
Availability details for
Quantity
  • Availability
  • Add to Cart