ADVH49B825DB AChemBlock

tert-butyl N-[(3S,4R)-3-methoxy-4-piperidyl]carbamate;oxalic acid

  • Molecular Weight : 320.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-3s-4r-3-methoxy-4-piperidyl-carbamate-oxalic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84470
Availability details for
Quantity
  • Availability
  • Add to Cart