ADVH49B825DF AChemBlock

tert-butyl N-[(3R,5R)-5-(hydroxymethyl)pyrrolidin-3-yl]-N-methyl-carbamate

  • Molecular Weight : 230.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-3r-5r-5-hydroxymethyl-pyrrolidin-3-yl-n-methyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84474
Availability details for
Quantity
  • Availability
  • Add to Cart