ADVH49B825E3 AChemBlock

tert-butyl N-[(3R,4R)-4-methylpyrrolidin-3-yl]carbamate hydrochloride

  • Molecular Weight : 236.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-3r-4r-4-methylpyrrolidin-3-yl-carbamate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84478
Availability details for
Quantity
  • Availability
  • Add to Cart