ADVH49B825E7 AChemBlock

tert-butyl N-[(2S,4R)-2-methylpiperidin-4-yl]carbamate

  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-2s-4r-2-methylpiperidin-4-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84483
Availability details for
Quantity
  • Availability
  • Add to Cart