ADVH49B825EC AChemBlock

2,2-dimethylcyclopropanecarboxamide

  • Cas Number : 1759-55-3
  • Molecular Weight : 113.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-2-dimethylcyclopropanecarboxamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78535
Availability details for
Quantity
  • Availability
  • Add to Cart