ADVH49B825EF AChemBlock

tert-butyl N-[(1S,3S)-rel-3-amino-3-methylcyclopentyl]carbamate

  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-1s-3s-rel-3-amino-3-methylcyclopentyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84491
Availability details for
Quantity
  • Availability
  • Add to Cart