ADVH49B825F1 AChemBlock

tert-butyl N-[(1S,2S,5R,6S)-rel-6-aminobicyclo[3.1.0]hexan-2-yl]carbamate

  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-1s-2s-5r-6s-rel-6-aminobicyclo-3-1-0-hexan-2-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84493
Availability details for
Quantity
  • Availability
  • Add to Cart