ADVH49B825F4 AChemBlock

tert-butyl N-[(1R,4r,6S)-6-hydroxyspiro[3.3]heptan-1-yl]carbamate

  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-1r-4r-6s-6-hydroxyspiro-3-3-heptan-1-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84496
Availability details for
Quantity
  • Availability
  • Add to Cart