ADVH49B825F6 AChemBlock

tert-butyl N-[(1R,3R)-rel-3-fluorocyclopentyl]carbamate

  • Molecular Weight : 203.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-1r-3r-rel-3-fluorocyclopentyl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84498
Availability details for
Quantity
  • Availability
  • Add to Cart