ADVH49B825F9 AChemBlock

tert-butyl N-[(1R)-3,3-difluoro-8-azaspiro[4.5]decan-1-yl]carbamate

  • Molecular Weight : 290.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-1r-3-3-difluoro-8-azaspiro-4-5-decan-1-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84501
Availability details for
Quantity
  • Availability
  • Add to Cart