ADVH49B825FD AChemBlock

bis(3,5-bis(trifluoromethyl)phenyl)methanone

  • Cas Number : 175136-66-0
  • Molecular Weight : 454.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/bis-3-5-bis-trifluoromethyl-phenyl-methanone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71623
Availability details for
Quantity
  • Availability
  • Add to Cart