ADVH49B82606 AChemBlock

(2R)-1,4-bis(tert-butoxycarbonyl)piperazine-2-carboxylic acid

  • Cas Number : 173774-48-6
  • Molecular Weight : 330.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2r-1-4-bis-tert-butoxycarbonyl-piperazine-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78583
Availability details for
Quantity
  • Availability
  • Add to Cart