ADVH49B82610 AChemBlock

tert-butyl N-(6-bromoindolizin-1-yl)carbamate

  • Molecular Weight : 311.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-6-bromoindolizin-1-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84524
Availability details for
Quantity
  • Availability
  • Add to Cart