ADVH49B82612 AChemBlock

(2-oxocyclobutyl) acetate

  • Cas Number : 17082-62-1
  • Molecular Weight : 128.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-oxocyclobutyl-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78628
Availability details for
Quantity
  • Availability
  • Add to Cart