ADVH49B82615 AChemBlock

3-(hydroxymethyl)piperidin-3-ol hydrochloride

  • Cas Number : 1706418-86-1
  • Molecular Weight : 167.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-hydroxymethyl-piperidin-3-ol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78677
Availability details for
Quantity
  • Availability
  • Add to Cart