ADVH49B8261A AChemBlock

3-chloro-5-iodo-isoquinoline

  • Cas Number : 1695831-18-5
  • Molecular Weight : 289.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-5-iodo-isoquinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78704
Availability details for
Quantity
  • Availability
  • Add to Cart